Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138893.1 | NP_620248.1 | 1, 2 | 272 | ||
XM_008771384.1 | XP_008769606.1 | 2, 3 | 1246 | |||
GenBank mRNA | AF209114.1 | - | 1, 2 | 272 | ||
AY004215.1 | - | 1, 2 | 272 |