Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270786.1 | NP_001257715.1 | 4 | 657 | ||
NM_001270787.1 | NP_001257716.1 | 4 | 903 | |||
NM_138904.2 | NP_620259.2 | 3 | 352 | |||
XM_006240728.2 | XP_006240790.1 | 4 | 683 | |||
XM_006240730.2 | XP_006240792.1 | 5 | 482 | |||
GenBank mRNA | BC089776.1 | - | 4 | 496 | ||
BC104712.1 | - | 4 | 401 | |||
FQ211735.1 | - | 4 | 903 | |||
J05499.1 | - | 3 | 351 |