Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190399.1 | NP_001177328.1 | 2 | 217 | ||
NM_138915.1 | NP_620270.1 | 2 | 276 | |||
GenBank mRNA | AF303658.1 | - | 2 | 276 | ||
FQ213422.1 | - | 2 | 191 |