- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Ss18l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138918.1 | NP_620273.1 | 4 | 257 | ||
GenBank mRNA | AY034073.1 | - | 4 | 257 |