Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130112.2 | NP_001123584.1 | 3, 4 | 375 | ||
NM_001206801.1 | NP_001193730.1 | 2, 3 | 249 | |||
NM_007370.5 | NP_031396.1 | 2, 3 | 249 | |||
NM_181578.3 | NP_853556.2 | 3, 4 | 613 | |||
XM_011538643.2 | XP_011536945.1 | 2, 3 | 247 | |||
XM_011538645.2 | XP_011536947.1 | 2, 3 | 247 | |||
XM_017019779.1 | XP_016875268.1 | 2, 3 | 247 | |||
GenBank mRNA | AB208992.1 | - | 2, 3 | 205 | ||
AK094575.1 | - | 3, 4 | 592 | |||
AK290685.1 | - | 2, 3 | 227 | |||
AK291107.1 | - | 2, 3 | 240 | |||
AU121576.1 | - | 3, 4 | 375 | |||
BC001866.1 | - | 2, 3 | 223 | 3544137 | ||
BC013961.2 | - | 2, 3 | 207 | 2960023 | ||
BC020440.1 | - | 2, 3 | 180 | |||
BM153733.1 | - | 2, 3 | 244 | |||
CR407651.1 | - | 2, 3 | 122 | |||
DB270923.1 | - | 2, 3 | 247 | |||
L07540.1 | - | 2, 3 | 131 |