Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139115.2 | NP_620815.2 | 6 | 833 | ||
XM_006246885.2 | XP_006246947.1 | 7 | 1910 | |||
XM_006246886.2 | XP_006246948.1 | 7 | 1912 | |||
XM_006246887.2 | XP_006246949.1 | 7 | 1910 | |||
XM_006246888.2 | XP_006246950.1 | 7 | 1912 | |||
XM_006246889.2 | XP_006246951.1 | 6 | 1790 | |||
XM_006246890.2 | XP_006246952.1 | 6 | 1790 | |||
XM_006246891.2 | XP_006246953.1 | 7 | 1911 | |||
GenBank mRNA | AF140359.1 | - | 7 | 1206 | ||
BC080241.1 | - | 6 | 2372 | |||
BC098656.1 | - | 6 | 833 | 7192854 |