Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trib3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144755.2 | NP_653356.1 | 4 | 1307 | ||
GenBank mRNA | AB020967.1 | - | 1292 | |||
BC091120.1 | - | 4 | 1307 | 7326024 |