Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Serpina4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145097.2 | NP_659565.2 | 2 | 533 | ||
XM_008764741.1 | XP_008762963.1 | 3 | 761 | |||
GenBank mRNA | BC088111.1 | - | 2 | 533 | 7368874 | |
FQ209558.1 | - | 2 | 550 | |||
FQ210636.1 | - | 2 | 553 | |||
FQ218781.1 | - | 1 | 301 | |||
FQ218835.1 | - | 2 | 553 | |||
FQ219263.1 | - | 1 | 301 | |||
FQ219393.1 | - | 2 | 603 | |||
FQ219554.1 | - | 1 | 301 | |||
U51017.1 | - | 1 | 494 |