Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145670.4 | NP_663703.3 | 11 | 2018 | ||
XM_008762466.1 | XP_008760688.1 | 12 | 2153 | |||
GenBank mRNA | AB029897.1 | - | 9 | 1493 | ||
AB180269.1 | - | 10 | 1661 | |||
AB180270.1 | - | 8 | 1427 | |||
AB180271.1 | - | 7 | 1385 | |||
AB180272.1 | - | 8 | 1358 | |||
BC094306.1 | - | 8 | 1415 | |||
BC103480.1 | - | 10 | 1916 |