Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153466.1 | NP_703196.1 | 5 | 655 | ||
GenBank mRNA | U57063.1 | - | 5 | 655 |
Gene Symbol: LOC100910060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770727.1 | XP_008768949.1 | 4 | 567 |