Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161521.1 | NP_001154993.1 | 4 | 504 | ||
NM_001161522.1 | NP_001154994.1 | 3 | 397 | |||
NM_007246.3 | NP_009177.3 | 4 | 627 | |||
XM_011531572.2 | XP_011529874.1 | 5 | 617 | |||
XM_017007674.1 | XP_016863163.1 | 5 | 577 | |||
XM_017007675.1 | XP_016863164.1 | 3 | 342 | |||
XM_017007676.1 | XP_016863165.1 | 3 | 450 | |||
GenBank mRNA | AF059569.1 | - | 4 | 476 | ||
AK025289.1 | - | 1 | 93 | |||
AK294103.1 | - | 4 | 498 | |||
AK315372.1 | - | 4 | 472 | |||
AL541403.3 | - | 4 | 627 | |||
BC022503.1 | - | 4 | 609 | 4791972 | ||
BC036468.2 | - | 4 | 568 | 4799622 | ||
BX647776.1 | - | 3 | 381 | |||
DA250099.1 | - | 4 | 504 |