Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gcnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173312.2 | NP_775434.1 | 2 | 415 | ||
XM_006243366.2 | XP_006243428.1 | 5 | 1110 | |||
XM_008766220.1 | XP_008764442.1 | 4 | 622 | |||
GenBank mRNA | AB098520.1 | - | 2 | 416 |