Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145036.1 | NP_001138508.1 | 13 | 1550 | ||
NM_007357.2 | NP_031383.1 | 13 | 1550 | |||
GenBank mRNA | AK295251.1 | - | 13 | 1557 | ||
AK302288.1 | - | 5 | 523 | |||
AK302791.1 | - | 14 | 1734 | |||
AK312678.1 | - | 13 | 1466 | |||
BC014960.1 | - | 13 | 1513 | 4876271 | ||
BC051906.1 | - | 13 | 1439 | 4541036 | ||
Z34975.1 | - | 13 | 1494 |