Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022623.2 | NP_072145.1 | 2 | 744 | ||
GenBank mRNA | AF183910.1 | - | 2 | 775 |
Set Membership: |
G-protein coupled receptor |