Gene Symbol: Inppl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270843.1 | NP_001257772.1 | 3 | 530 | ||
NM_022944.2 | NP_075233.1 | 4 | 586 | |||
GenBank mRNA | AB011439.1 | - | 3 | 530 | ||
AB025794.1 | - | 4 | 584 | |||
FQ139232.1 | - | 4 | 622 |