Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024154.2 | NP_077068.1 | 3 | 574 | ||
XM_006257381.2 | XP_006257443.1 | 3 | 859 | |||
XM_008765794.1 | XP_008764016.1 | 3 | 859 | |||
XM_008765795.1 | XP_008764017.1 | 3 | 512 | |||
XM_008765796.1 | XP_008764018.1 | 3 | 859 |