Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031011.3 | NP_112273.3 | 2, 3 | 518 | ||
GenBank mRNA | AY724509.1 | - | 2, 3 | 201 | ||
BC061532.1 | - | 2, 3 | 521 | 6921099 | ||
FM031711.1 | - | 2, 3 | 165 | |||
FQ232080.1 | - | 2, 3 | 338 | |||
FQ234346.1 | - | 2, 3 | 341 | |||
M34464.1 | - | 2, 3 | 464 |