Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031011.3 | NP_112273.3 | 8 | 1513 | ||
GenBank mRNA | AY724509.1 | - | 8 | 1196 | ||
BC061532.1 | - | 8 | 1516 | 6921099 | ||
M34464.1 | - | 8 | 1459 |