Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031081.1 | NP_112343.1 | 4, 5 | 528 | ||
XM_006246036.2 | XP_006246098.1 | 4, 5 | 2380 | |||
XM_006246038.2 | XP_006246100.1 | 4, 5 | 590 | |||
GenBank mRNA | Y15748.1 | - | 4, 5 | 528 |