Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031106.1 | NP_112368.1 | 2 | 126 | ||
GenBank mRNA | BC059132.1 | - | 2 | 123 | 6919306 | |
BC069173.1 | - | 2 | 108 | 7103990 | ||
FQ210660.1 | - | 2 | 129 | |||
FQ210874.1 | - | 2 | 129 | |||
FQ217102.1 | - | 2 | 129 | |||
FQ217508.1 | - | 2 | 119 | |||
FQ220738.1 | - | 2 | 129 | |||
FQ221496.1 | - | 2 | 119 | |||
FQ221700.1 | - | 2 | 119 | |||
FQ222983.1 | - | 2 | 129 | |||
FQ223019.1 | - | 2 | 129 | |||
FQ223296.1 | - | 2 | 134 | |||
FQ223471.1 | - | 2 | 119 | |||
S79981.1 | - | 2 | 126 | |||
X66369.1 | - | 2 | 126 |
Gene Symbol: LOC100360841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730037.4 | XP_002730083.1 | 1 | 122 |
Gene Symbol: RGD1561310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771609.1 | XP_008769831.1 | 1 | 122 | ||
XM_574022.5 | XP_574022.3 | 1 | 122 |