Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281425.1 | NP_001268354.1 | 6 | 625 | ||
NM_001281426.1 | NP_001268355.1 | 7 | 740 | |||
NM_001281427.1 | NP_001268356.1 | 6 | 679 | |||
NM_007234.4 | NP_009165.1 | 7 | 709 | |||
NM_024348.3 | NP_077324.1 | 7 | 806 | |||
GenBank mRNA | AF082513.1 | - | 7 | 666 | ||
AK055199.1 | - | 2 | 2881 | |||
AK091661.1 | - | 5 | 3736 | |||
BC000319.2 | - | 7 | 757 | 2821720 | ||
BC003004.1 | - | 7 | 757 | 2821720 | ||
BC107697.1 | - | 7 | 678 | 6454606 | ||
BG715167.1 | - | 7 | 690 | |||
CN257020.1 | - | 5 | 461 |