Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161521.1 | NP_001154993.1 | 15 | 2325 | ||
NM_001161522.1 | NP_001154994.1 | 14 | 2218 | |||
NM_007246.3 | NP_009177.3 | 15 | 2448 | |||
XM_011531572.2 | XP_011529874.1 | 16 | 2438 | |||
XM_011531575.2 | XP_011529877.1 | 14 | 2270 | |||
XM_011531576.2 | XP_011529878.1 | 13 | 1938 | |||
XM_017007674.1 | XP_016863163.1 | 16 | 2398 | |||
XM_017007676.1 | XP_016863165.1 | 14 | 2271 | |||
XM_017007677.1 | XP_016863166.1 | 13 | 2145 | |||
GenBank mRNA | AB209727.1 | - | 11 | 1792 | ||
AF059569.1 | - | 15 | 2297 | |||
AK025289.1 | - | 12 | 1914 | |||
AK308940.1 | - | 11 | 1694 | |||
BC022503.1 | - | 15 | 2430 | 4791972 | ||
BC036468.2 | - | 15 | 2389 | 4799622 | ||
BX647776.1 | - | 14 | 2202 |