Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031719.1 | NP_113907.1 | 3, 4 | 406 | ||
XM_006229792.2 | XP_006229854.1 | 3, 4 | 469 | |||
GenBank mRNA | BC060555.1 | - | 3, 4 | 450 | 6918578 | |
D13985.1 | - | 3, 4 | 476 | |||
D17698.1 | - | 3, 4 | 389 | |||
FQ212666.1 | - | 1, 2 | 88 | |||
FQ232880.1 | - | 1, 2 | 88 | |||
L26450.1 | - | 3, 4 | 406 |