Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135752.1 | NP_001129224.1 | 3 | 509 | ||
NM_001135753.1 | NP_001129225.1 | 3 | 509 | |||
NM_007265.2 | NP_009196.1 | 3 | 509 | |||
NR_024203.1 | 2 | 291 | ||||
GenBank mRNA | AK022616.1 | - | 3 | 457 | ||
AK225519.1 | - | 3 | 485 | |||
AK315711.1 | - | 3 | 482 | |||
BC000721.2 | - | 3 | 473 | 3504956 | ||
BM763002.1 | - | 2 | 292 | |||
CR457407.1 | - | 2 | 266 | |||
D88208.1 | - | 3 | 473 | |||
DA162516.1 | - | 3 | 485 |