Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc16a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031744.2 | NP_113932.2 | 5 | 897 | ||
XM_006242039.2 | XP_006242101.1 | 4 | 1166 | |||
GenBank mRNA | AF059258.1 | - | 5 | 897 |