Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103692171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763086.1 | XP_008761308.1 | 2727 |
Gene Symbol: Gcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031749.2 | NP_113937.1 | 4 | 2659 | ||
GenBank mRNA | AF087431.1 | - | 4 | 2660 |