Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053348.1 | NP_445800.1 | 6 | 832 | ||
XM_006248549.1 | XP_006248611.1 | 7 | 786 | |||
XM_006248550.1 | XP_006248612.1 | 7 | 1149 | |||
GenBank mRNA | AJ242926.1 | - | 6 | 832 | ||
BC070508.1 | - | 7 | 771 | 7099154 | ||
FQ209398.1 | - | 6 | 844 | |||
FQ209449.1 | - | 7 | 769 | |||
FQ209518.1 | - | 6 | 783 | |||
FQ209610.1 | - | 6 | 819 | |||
FQ209832.1 | - | 6 | 784 | |||
FQ210294.1 | - | 6 | 783 | |||
FQ210335.1 | - | 6 | 784 | |||
FQ211050.1 | - | 6 | 783 | |||
FQ211248.1 | - | 6 | 818 | |||
FQ218476.1 | - | 6 | 814 | |||
FQ218884.1 | - | 7 | 754 | |||
FQ218893.1 | - | 5 | 684 | |||
FQ218905.1 | - | 6 | 783 | |||
FQ218930.1 | - | 6 | 816 | |||
FQ219048.1 | - | 6 | 814 | |||
FQ219152.1 | - | 6 | 820 | |||
FQ219190.1 | - | 6 | 819 | |||
FQ219266.1 | - | 6 | 783 | |||
FQ219352.1 | - | 6 | 783 | |||
FQ219426.1 | - | 4 | 385 | |||
FQ219454.1 | - | 6 | 783 | |||
FQ219459.1 | - | 6 | 781 | |||
FQ219500.1 | - | 6 | 786 | |||
FQ219513.1 | - | 6 | 855 |