Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053360.2 | NP_445812.1 | 2 | 408 | ||
XM_006256931.1 | XP_006256993.1 | 2 | 569 | |||
GenBank mRNA | AF230519.1 | - | 2 | 220 | ||
AF255888.1 | - | 2 | 326 | |||
BC070877.1 | - | 2 | 408 | 7096160 |