Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053424.1 | NP_445876.1 | 4 | 382 | ||
XM_006250788.2 | XP_006250850.1 | 4 | 842 | |||
XM_006250789.2 | XP_006250851.1 | 4 | 842 | |||
XM_006250790.2 | XP_006250852.1 | 5 | 560 | |||
XM_006250791.2 | XP_006250853.1 | 4 | 394 | |||
GenBank mRNA | AF004017.1 | - | 1 | 220 | ||
AF027362.1 | - | 1 | 252 | |||
AF124441.1 | - | 4 | 382 | |||
AF210250.1 | - | 4 | 389 | |||
AF254802.1 | - | 4 | 382 |