Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053546.1 | NP_445998.1 | 1 | 383 | ||
XM_006241609.2 | XP_006241671.1 | 1 | 725 | |||
GenBank mRNA | AB080023.1 | - | 1 | 383 |