Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012071.3 | NP_036203.1 | 8 | 889 | ||
GenBank mRNA | AF078848.1 | - | 8 | 864 | ||
AF201948.1 | - | 8 | 883 | |||
AK128789.1 | - | 5 | 1404 | |||
BC022898.2 | - | 8 | 876 | 3531636 | ||
BX647275.1 | - | 5 | 2248 |