Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053646.1 | NP_446098.1 | 4 | 490 | ||
XM_006231261.2 | XP_006231323.1 | 7 | 848 | |||
XM_006231263.1 | XP_006231325.1 | 6 | 1426 | |||
XM_006231267.2 | XP_006231329.1 | 6 | 1432 | |||
XM_006231268.2 | XP_006231330.1 | 6 | 1429 | |||
XM_006231270.1 | XP_006231332.1 | 6 | 1429 | |||
XM_008760319.1 | XP_008758541.1 | 9 | 3268 | |||
XM_008760320.1 | XP_008758542.1 | 6 | 709 | |||
GenBank mRNA | AB057433.1 | - | 4 | 490 |