Search Thermo Fisher Scientific
Gene Symbol: Ntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053731.2 | NP_446183.2 | 2 | 1073 | ||
XM_006246567.2 | XP_006246629.1 | 3 | 1425 | |||
XM_006246568.2 | XP_006246630.1 | 3 | 1259 | |||
GenBank mRNA | AY028417.1 | - | 2 | 1073 | ||
Y11353.1 | - | 2 | 113 |