Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053742.1 | NP_446194.1 | 3, 4 | 221 | ||
XM_006249522.2 | XP_006249584.1 | 3, 4 | 265 | |||
XM_006249523.2 | XP_006249585.1 | 3, 4 | 265 | |||
GenBank mRNA | BC061538.1 | - | 3, 4 | 253 | 6888321 | |
D21132.1 | - | 3, 4 | 221 |