Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053821.2 | NP_446273.1 | 4 | 569 | ||
GenBank mRNA | BC072505.1 | - | 4 | 569 | 7099019 | |
L19699.1 | - | 3 | 417 |