Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053821.2 | NP_446273.1 | 5 | 1435 | ||
GenBank mRNA | BC072505.1 | - | 5 | 1435 | 7099019 | |
L19699.1 | - | 4 | 1292 |