Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: APPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012096.2 | NP_036228.1 | 4 | 384 | ||
XM_011533583.2 | XP_011531885.1 | 5 | 437 | |||
GenBank mRNA | AB037849.2 | - | 4 | 237 | ||
AF169797.1 | - | 4 | 295 | |||
AF424738.1 | - | 4 | 293 | |||
AK299008.1 | - | 5 | 426 | |||
AK310716.1 | - | 4 | 332 | |||
BC028599.2 | - | 4 | 384 | 4829430 |