Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHORDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012124.2 | NP_036256.2 | 2, 3 | 532 | ||
XM_017017540.1 | XP_016873029.1 | 2, 3 | 245 | |||
XM_017017541.1 | XP_016873030.1 | 2, 3 | 246 | |||
GenBank mRNA | AF192466.1 | - | 2, 3 | 206 | ||
AK225365.1 | - | 2, 3 | 234 | |||
AK225565.1 | - | 2, 3 | 217 | |||
AK226099.1 | - | 2, 3 | 209 | |||
AK290231.1 | - | 2, 3 | 532 | |||
AK312663.1 | - | 2, 3 | 217 | |||
BC072461.1 | - | 2, 3 | 181 | 6471725 | ||
DA417105.1 | - | 2, 3 | 532 |
Gene Symbol: LOC727896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026659.1 | 1 | 209 |