Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130779.2 | NP_570135.2 | 3, 4 | 1268 | ||
XM_006239847.2 | XP_006239909.1 | 2, 3 | 1004 | |||
XM_006239848.2 | XP_006239910.1 | 2, 3 | 1007 | |||
XM_006239849.2 | XP_006239911.1 | 2, 3 | 1001 | |||
XM_006239850.2 | XP_006239912.1 | 2, 3 | 1010 | |||
XM_008764532.1 | XP_008762754.1 | 3, 4 | 1124 | |||
GenBank mRNA | M55075.1 | - | 3, 4 | 1193 |