Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133300.3 | NP_579834.2 | 5 | 760 | ||
GenBank mRNA | BC080243.1 | - | 5 | 709 | 7100155 | |
FQ212901.1 | - | 5 | 746 | |||
M75168.1 | - | 4 | 665 |