Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133300.3 | NP_579834.2 | 6 | 868 | ||
GenBank mRNA | BC080243.1 | - | 6 | 817 | 7100155 | |
M75168.1 | - | 5 | 773 |