Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou3f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138838.1 | NP_620193.1 | 1 | 2095 | ||
GenBank mRNA | M72711.1 | - | 1 | 2095 |