Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139114.3 | NP_620814.1 | 2, 3 | 255 | ||
GenBank mRNA | AA687059.1 | - | 2, 3 | 260 | ||
BC078724.1 | - | 1, 2 | 194 | 7125608 | ||
FQ210520.1 | - | 2, 3 | 210 | |||
FQ217417.1 | - | 2, 3 | 211 | |||
FQ218767.1 | - | 2, 3 | 210 | |||
FQ221301.1 | - | 2, 3 | 223 | |||
FQ221436.1 | - | 2, 3 | 212 | |||
FQ221541.1 | - | 2, 3 | 211 | |||
FQ221627.1 | - | 2, 3 | 214 | |||
FQ221775.1 | - | 2, 3 | 211 | |||
FQ221788.1 | - | 2, 3 | 210 | |||
FQ221816.1 | - | 2, 3 | 211 | |||
FQ222173.1 | - | 2, 3 | 211 | |||
FQ222227.1 | - | 2, 3 | 214 | |||
FQ222322.1 | - | 2, 3 | 210 | |||
FQ222450.1 | - | 2, 3 | 224 | |||
FQ222725.1 | - | 2, 3 | 211 | |||
FQ224390.1 | - | 2, 3 | 210 | |||
FQ227511.1 | - | 2, 3 | 225 | |||
FQ228660.1 | - | 1, 2 | 199 | |||
FQ230911.1 | - | 2, 3 | 214 | |||
X78167.1 | - | 1, 2 | 192 |
Gene Symbol: RGD1565131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248902.2 | XP_006248964.1 | 1 | 206 | ||
XM_008769031.1 | XP_008767253.1 | 1 | 206 |
Gene Symbol: RGD1565767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230013.6 | XP_230013.1 | 1 | 183 |