Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153730.2 | NP_714952.1 | 3 | 558 | ||
XM_006249843.2 | XP_006249905.1 | 4 | 707 | |||
XM_006249844.2 | XP_006249906.1 | 4 | 981 | |||
XM_006249845.1 | XP_006249907.1 | 4 | 687 | |||
XM_006249846.2 | XP_006249908.1 | 5 | 773 | |||
XM_008769539.1 | XP_008767761.1 | 4 | 746 | |||
XM_008769540.1 | XP_008767762.1 | 5 | 601 | |||
XM_008769541.1 | XP_008767763.1 | 4 | 491 | |||
GenBank mRNA | AJ133126.1 | - | 3 | 600 | ||
BC086983.1 | - | 4 | 688 | 7133597 | ||
FQ210179.1 | - | 5 | 757 |