Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172035.1 | NP_742032.1 | 1 | 278 | ||
GenBank mRNA | L02530.1 | - | 1 | 278 |
Set Membership: |
![]() |