Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772679.1 | XP_008770901.1 | 6 | 1809 | ||
GenBank mRNA | AY228140.1 | - | 4 | 594 | ||
BC081694.1 | - | 4 | 838 |
Gene Symbol: LOC690182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759848.1 | XP_008758070.1 | 1 | 834 |