Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181772.1 | NP_861437.1 | 3 | 367 | ||
GenBank mRNA | AY288431.1 | - | 3 | 367 |
Set Membership: |
G-protein coupled receptor |