Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198131.1 | NP_937764.1 | 2 | 304 | ||
GenBank mRNA | AY191271.1 | - | 2 | 304 | ||
AY859725.1 | - | 1 | 208 |