Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198742.2 | NP_942037.2 | 12 | 1807 | ||
XM_006232507.1 | XP_006232569.1 | 11 | 1660 | |||
XM_008761084.1 | XP_008759306.1 | 13 | 2033 | |||
GenBank mRNA | AY365022.1 | - | 12 | 1633 | ||
BC081890.1 | - | 12 | 1807 | 7109511 | ||
FQ209791.1 | - | 6 | 836 | |||
FQ211230.1 | - | 12 | 1864 | |||
FQ218772.1 | - | 14 | 2174 |