Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005458.7 | NP_005449.5 | 18 | 3016 | ||
XM_005252316.4 | XP_005252373.1 | 16 | 2049 | |||
XM_017015331.1 | XP_016870820.1 | 17 | 2434 | |||
XM_017015332.1 | XP_016870821.1 | 15 | 2003 | |||
GenBank mRNA | AF056085.1 | - | 18 | 3013 | ||
AF069755.1 | - | 18 | 2712 | |||
AF074483.1 | - | 18 | 2556 | |||
AF095784.1 | - | 18 | 2687 | |||
AF099033.1 | - | 18 | 2556 | |||
AJ012188.1 | - | 18 | 2556 | |||
AK308139.1 | - | 17 | 2187 | |||
BC035071.2 | - | 19 | 2851 | 5260138 | ||
JQ308173.1 | - | 18 | 2556 |
Set Membership: |
![]() |